Learn More
Cayman Chemical FFN-102 (trifluoroacetate salt), Purity: ≥98%, CAS: 1234064-11-9, 10 mg
SDP
Catalog No. 502994000
Encompass_Preferred
A pH-dependent fluorescent probe to label dopaminergic cells; excitation spectra of 340 and 370 nm at pH 5 and 7.4, respectively, which correspond to vesicular and cytoplasmic pH values; emission spectrum of 453 nm, with a higher intensity at pH 7.4; inhibits DAT (13.6% at a concentration of 10 µM) and the 5-HT receptor subtype 5-HT2c (Ki = ~3 µM); does not bind to 37 central nervous system receptors, including dopamine and serotonin receptors, up to a concentration of 10 µM
Catalog No. 50-299-4000 Supplier Cayman Chemical Supplier No. 2145810 MG
A pH-dependent fluorescent probe to label dopaminergic cells; excitation spectra of 340 and 370 nm at pH 5 and 7.4, respectively, which correspond to vesicular and cytoplasmic pH values; emission spectrum of 453 nm, with a higher intensity at pH 7.4; inhibits DAT (13.6% at a concentration of 10 µM) and the 5-HT receptor subtype 5-HT2c (Ki = ~3 µM); does not bind to 37 central nervous system receptors, including dopamine and serotonin receptors, up to a concentration of 10 µM