Learn More
Abcam FFN202 (Mini 202), Fluorescent VMAT1 and VMAT2 substrate, 5MG
Catalog No. 502707185
Encompass_Preferred
MW 353.68 g/mol, Purity >97%. Novel, pH responsive fluorescent false neurotransmitter (FFN). Rodent VMAT1 and VMAT2 substrate. Enables in situ pH measurement of secretory catecholamine vesicles in PC-12 cells. Sufficiently bright, photostable and suitable for two-photon fluorescence microscopy and standard fluorescent microscopy. Compatible with GFP tags.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7185 Supplier Abcam Supplier No. AB1208675MG
Add to Cart
Add to Cart
MW 353.68 g/mol, Purity >97%. Novel, pH responsive fluorescent false neurotransmitter (FFN). Rodent VMAT1 and VMAT2 substrate. Enables in situ pH measurement of secretory catecholamine vesicles in PC-12 cells. Sufficiently bright, photostable and suitable for two-photon fluorescence microscopy and standard fluorescent microscopy. Compatible with GFP tags.
The product is subject to the following: Abcam Restricted Use Statement