Learn More
Medchemexpress LLC FFN270 hydrochloride | 2341841-05-0 | 99.0% | 259.66 g/mol | C11H11ClFNO3 | 5 MG
SDP
Catalog No. 5000262139
Encompass_Preferred
FFN270 hydrochloride is a fluorescent tracer of norepinephrine used as a fluorescent substrate for norepinephrine transporters (NET) and vesicular monoamine transporters (VMAT). Provided as the hydrochloride salt, it is intended for biochemical and cellular assays to monitor norepinephrine uptake and vesicular transport.
  • Fluorescent tracer for norepinephrine transport studies.
  • Acts as a substrate for NET and VMAT, enabling uptake and vesicular labeling.
  • High purity (≥99.0%) suitable for research applications.
  • White to off-white solid with molecular weight 259.66 g/mol and formula C11H11ClFNO3.
  • Storage recommendations: sealed, protected from moisture; in solvent -80°C (6 months), -20°C (1 month).
  • Available in small laboratory quantities for assay development and validation.
Catalog No. 50-002-62139 Supplier Medchemexpress LLC Supplier No. HY1310075MG
FFN270 hydrochloride is a fluorescent tracer of norepinephrine used as a fluorescent substrate for norepinephrine transporters (NET) and vesicular monoamine transporters (VMAT). Provided as the hydrochloride salt, it is intended for biochemical and cellular assays to monitor norepinephrine uptake and vesicular transport.
  • Fluorescent tracer for norepinephrine transport studies.
  • Acts as a substrate for NET and VMAT, enabling uptake and vesicular labeling.
  • High purity (≥99.0%) suitable for research applications.
  • White to off-white solid with molecular weight 259.66 g/mol and formula C11H11ClFNO3.
  • Storage recommendations: sealed, protected from moisture; in solvent -80°C (6 months), -20°C (1 month).
  • Available in small laboratory quantities for assay development and validation.