Learn More
Abcam FFN511, Fluorescent substrate for VMAT2, 1MG
Catalog No. 502707670
Encompass_Preferred
MW 284.35 Da, Purity >98%. First novel fluorescent false neurotransmitter. Enables optical imaging of presynaptic terminal activity. Inhibits 5-HT binding to VMAT2 (IC₅₀ = 1 μM (comparable to dopamine itself)). Released via exocytosis in the striatum in mouse acute slice preparation. Labels dopamine and other presynaptic terminals in the striatum. Compatible with GFP tags and other optical probes. Sufficiently bright, photostable and suitable for two-photon fluorescence microscopy. View the technique online For highly selective labelling of dopaminergic presynaptic termainals FFN102 (ab120866) is recommended.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7670 Supplier Abcam Supplier No. AB1203311MG
MW 284.35 Da, Purity >98%. First novel fluorescent false neurotransmitter. Enables optical imaging of presynaptic terminal activity. Inhibits 5-HT binding to VMAT2 (IC₅₀ = 1 μM (comparable to dopamine itself)). Released via exocytosis in the striatum in mouse acute slice preparation. Labels dopamine and other presynaptic terminals in the striatum. Compatible with GFP tags and other optical probes. Sufficiently bright, photostable and suitable for two-photon fluorescence microscopy. View the technique online For highly selective labelling of dopaminergic presynaptic termainals FFN102 (ab120866) is recommended.
The product is subject to the following: Abcam Restricted Use Statement