Learn More
Biochain Institute Inc FFPE and Frozen Matched Pair Total RNA: Human Adult Normal Tissue: Liver, 2 x 1 ug/pk
SDP
Catalog No. 501871554
Encompass_Preferred
The same normal tissue is collected with both FFPE and Snap Frozen format at the same time. Comparing the qRT-PCR or next generation sequencing data with total RNA isolated from both tissue format will help scientists to develope diagnostic method from FFPE samples, which are major clinical tissue sample
Catalog No. 50-187-1554 Supplier Biochain Institute Inc Supplier No. R8234149FP
The same normal tissue is collected with both FFPE and Snap Frozen format at the same time. Comparing the qRT-PCR or next generation sequencing data with total RNA isolated from both tissue format will help scientists to develope diagnostic method from FFPE samples, which are major clinical tissue sample