Learn More
Biochain Institute Inc FFPE Genomic DNA - Human Tumor Tissue: Uterus, 2 ug/pk
SDP
Catalog No. 501871520
Encompass_Preferred
FFPE Genomic DNA FFPE DNAs are isolated from Formalin Fixed Paraffin Embedded mater. More cancer/normal FFPE DNA samples for NGS and PCR applications are available.
Catalog No. 50-187-1520 Supplier Biochain Institute Inc Supplier No. D2235274
FFPE Genomic DNA FFPE DNAs are isolated from Formalin Fixed Paraffin Embedded mater. More cancer/normal FFPE DNA samples for NGS and PCR applications are available.