Learn More
CD BIOSYSTEMS INC FFPE LIBRARY AUTOMATION KIT
Catalog No. NC3360364
Encompass_Preferred
  • Engineered for streamlined library preparation from FFPE and fixed probe-based samples.
  • Offers an automated solution to enhance reproducibility and efficiency in library construction.
  • Includes essential modules for gene expression analysis.
  • Incorporates sample integrity tracking oligos for reliable experimental outcomes.
  • Compatible with the MGI SP-100 platform.
  • Has a reaction capacity of 16 reactions.
  • Features a gene expression library module.
  • Designed for user-friendly operation and consistency.
Catalog No. NC3360364 Supplier CD BIOSYSTEMS INC Supplier No. FP400
Add to Cart
Add to Cart
Request a Quote
  • Engineered for streamlined library preparation from FFPE and fixed probe-based samples.
  • Offers an automated solution to enhance reproducibility and efficiency in library construction.
  • Includes essential modules for gene expression analysis.
  • Incorporates sample integrity tracking oligos for reliable experimental outcomes.
  • Compatible with the MGI SP-100 platform.
  • Has a reaction capacity of 16 reactions.
  • Features a gene expression library module.
  • Designed for user-friendly operation and consistency.