Learn More
Bioworld FFPE RNA/DNA PURIFICATION KIT
Catalog No. 50196926
Encompass_Preferred
For the rapid isolation and purification of total RNA (including microRNA) and genomic DNA from FFPE tissue samples. Purify all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA); ...
Catalog No. 50-196-926 Supplier Bioworld Supplier No. 107600711
For the rapid isolation and purification of total RNA (including microRNA) and genomic DNA from FFPE tissue samples. Purify all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA); ...