Learn More
Norgen Biotek Corp FFPE RNA Purification Kit
Catalog No. NC1579384
Encompass_Preferred
Norgen’s FFPE RNA Purification Kit provides a rapid method for the isolation and purification of total RNA (including microRNA) from formalin-fixed paraffin-embedded (FFPE) tissue samples in as little as 1 hour. Using formalin to fix tissues leads to crosslinking of the RNA and proteins, and the process of embedding the tissue samples can also lead to fragmentation of the RNA over time. Norgen’s FFPE RNA Purification Kit provides conditions that allow for the partial reversing of the formalin modifications, resulting in a high quality and yield of RNA. The kit is able to purify all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA), depending on the age of the FFPE tissue as fragmentation of the RNA is known to occur over time.
Catalog No. NC1579384 Supplier Norgen Biotek Corp Supplier No. 25300
Norgen’s FFPE RNA Purification Kit provides a rapid method for the isolation and purification of total RNA (including microRNA) from formalin-fixed paraffin-embedded (FFPE) tissue samples in as little as 1 hour. Using formalin to fix tissues leads to crosslinking of the RNA and proteins, and the process of embedding the tissue samples can also lead to fragmentation of the RNA over time. Norgen’s FFPE RNA Purification Kit provides conditions that allow for the partial reversing of the formalin modifications, resulting in a high quality and yield of RNA. The kit is able to purify all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA), depending on the age of the FFPE tissue as fragmentation of the RNA is known to occur over time.