Learn More
Biochain Institute Inc FFPE Tissue DNA Extraction Kit - Column, 1 kit/pk
SDP
Supplier:  Biochain Institute Inc K5011350
Encompass_Preferred
BioChain’s FFPE Tissue DNA Extraction Kit - Column allows for facile and efficient deoxyribonucleic acid extraction from FFPE tissues, with potential high throughput capabilities and full compatibility for downstream applications such as qPCR. Utilizing heat and proteinase K treatment, BioChain’s FFPE Tissue DNA Extraction Kit- Column is optimized in the removal of paraffin, partial reversal of formalin cross linking, and release of DNA from fixed tissues. After clean up with DNA binding columns, the concentration of the high purity DNA can be determined by spectrophotometer or nanodrop equipment.
Catalog No. 50-187-1529