Learn More
Biochain Institute Inc FFPE Tissue DNA Extraction Kit — Magnetic Beads, 1 kit/pk
SDP
Catalog No. 501871530
Encompass_Preferred
BioChain’s FFPE Tissue DNA Extraction Kit - Magnetic Beads allows for facile and efficient deoxyribonucleic acid extraction from FFPE tissues, with potential high throughput capabilities and full compatibility for down-stream applications such as qPCR. Utilizing heat and proteinase K treatment, BioChain’s FFPE Tissue DNA Extraction Kit- Magnetic beads is optimized in the removal of paraffin, artial reversal of formalin cross linking, and release of DNA from fixed tissues. After clean up with DNA inding beads, the concentration of the high urity DNA can be determined by spectrophotometer or anodrop equipment.
Catalog No. 50-187-1530 Supplier Biochain Institute Inc Supplier No. K5011450
BioChain’s FFPE Tissue DNA Extraction Kit - Magnetic Beads allows for facile and efficient deoxyribonucleic acid extraction from FFPE tissues, with potential high throughput capabilities and full compatibility for down-stream applications such as qPCR. Utilizing heat and proteinase K treatment, BioChain’s FFPE Tissue DNA Extraction Kit- Magnetic beads is optimized in the removal of paraffin, artial reversal of formalin cross linking, and release of DNA from fixed tissues. After clean up with DNA inding beads, the concentration of the high urity DNA can be determined by spectrophotometer or anodrop equipment.