Learn More
TARGETMOL CHEMICALS INC FG-2216 25MG
SDP
Catalog No. 502580805
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FG-2216 (YM-311) is a potent HIF-prolyl hydroxylase inhibitor for the PDH2 enzyme (IC50 3.9 uM); orally bioavailable and induced reversible and significant Epo induction in vivo. purity: 99%
Catalog No. 50-258-0805 Supplier TARGETMOL CHEMICALS INC Supplier No. T244525MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FG-2216 (YM-311) is a potent HIF-prolyl hydroxylase inhibitor for the PDH2 enzyme (IC50 3.9 uM); orally bioavailable and induced reversible and significant Epo induction in vivo. purity: 99%