Learn More
Selleck Chemical LLC FG-2216
SDP
Catalog No. 5000000472
Encompass_Preferred
FG-2216 is a potent and orally active HIF prolyl 4-hydroxylase inhibitor with IC50 of 3 9 M for PHD2 Phase 2
Catalog No. 50-000-00472 Supplier Selleck Chemical LLC Supplier No. S797925MG
FG-2216 is a potent and orally active HIF prolyl 4-hydroxylase inhibitor with IC50 of 3 9 M for PHD2 Phase 2