Learn More
Medchemexpress LLC HY-15641 5mg Medchemexpress, FG-2216 CAS:223387-75-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY156415MG
Encompass_Preferred
Medchemexpress, HY-15641 5mg FG-2216 CAS:223387-75-5 FG-2216 (IOX3) is a potent and orally active inhibitor of HIF prolyl hydroxylase-2 (PHD2), with an IC50 of 3.9 nM. FG-2216 induces robust erythropoietin and modest fetal hemoglobin in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1762