Learn More
Medchemexpress LLC FG-2216 | 859353-53-8 | MFCD00672101 | >98% | 260.27 | 10 MG
SDP
Catalog No. 5000365133
Encompass_Preferred
FG-2216 (IOX3) is a potent and orally active inhibitor of HIF prolyl hydroxylase-2 (PHD2). It induces robust erythropoietin and modest fetal hemoglobin in vivo.
  • Potent and orally active inhibitor of HIF prolyl hydroxylase-2 (PHD2)
  • IC50 of 3.9 μM against PHD2
  • Induces robust erythropoietin in vivo
  • Induces modest fetal hemoglobin in vivo
  • Stabilizes HIF-1α and HIF-2α in Hep3B cells
Catalog No. 50-003-65133 Supplier Medchemexpress LLC Supplier No. 10MGHY15641
FG-2216 (IOX3) is a potent and orally active inhibitor of HIF prolyl hydroxylase-2 (PHD2). It induces robust erythropoietin and modest fetal hemoglobin in vivo.
  • Potent and orally active inhibitor of HIF prolyl hydroxylase-2 (PHD2)
  • IC50 of 3.9 μM against PHD2
  • Induces robust erythropoietin in vivo
  • Induces modest fetal hemoglobin in vivo
  • Stabilizes HIF-1α and HIF-2α in Hep3B cells