Learn More
Medchemexpress LLC FG-3019 1mg | 1mg
SDP
Catalog No. 5000324099
Encompass_Preferred
FG-3019 (Pamrevlumab) is a recombinant human antibody that binds to connective tissue growth factor (CTGF) FG-3019 can be used for the research of idiopathic pulmonary fibrosis (IPF)[1][2]
Catalog No. 50-003-24099 Supplier Medchemexpress LLC Supplier No. HYP992881MG
FG-3019 (Pamrevlumab) is a recombinant human antibody that binds to connective tissue growth factor (CTGF) FG-3019 can be used for the research of idiopathic pulmonary fibrosis (IPF)[1][2]