Learn More
Medchemexpress LLC FG-3019 5mg | 5mg
SDP
Catalog No. 5000323661
Encompass_Preferred
FG-3019 (Pamrevlumab) is a recombinant human antibody that binds to connective tissue growth factor (CTGF) FG-3019 can be used for the research of idiopathic pulmonary fibrosis (IPF)[1][2]
Catalog No. 50-003-23661 Supplier Medchemexpress LLC Supplier No. HYP992885MG
FG-3019 (Pamrevlumab) is a recombinant human antibody that binds to connective tissue growth factor (CTGF) FG-3019 can be used for the research of idiopathic pulmonary fibrosis (IPF)[1][2]