Learn More
Medchemexpress LLC FG-3246 FOR46 1mg | 1mg
SDP
Catalog No. 5000324857
Encompass_Preferred
FG-3246 (FOR46) is a humanized antibody expressed in CHO cells that targets CD46 FG-3246 (FOR46) has a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 143 6 kDa The isotype control for FG-3246 (FOR46) can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24857 Supplier Medchemexpress LLC Supplier No. HYP9905861MG
FG-3246 (FOR46) is a humanized antibody expressed in CHO cells that targets CD46 FG-3246 (FOR46) has a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 143 6 kDa The isotype control for FG-3246 (FOR46) can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)