Learn More
Apexbio Technology LLC FG-4592 (ASP1517), 50mg. Cas: 808118-40-3 MFCD: MFCD20040519. HIF prolyl-hydroxylase inhibitor
SDP
Supplier:  Apexbio Technology LLC A418750
Encompass_Preferred
FG-4592 (also known as ASP1517), 2-(4-hydroxy-1-methyl-7-phenoxyisoquinoline-3-carboxamido)acetic acid, is a potent small molecule inhibitor of hypo (Epo). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4902