Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ 96-Well Support Base

Catalog No. 4379590
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 bases
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-795-90 10 bases
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-795-90 Supplier Applied Biosystems™ Supplier No. 4379590
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Contains 10 MicroAmp 96-Well Support Bases

Designed to hold MicroAmp Reaction Tubes in Tray/Retainer Sets or MicroAmp 48- and 96-Well Plates for convenient use in pre- and post-PCR sample processing

  • Convenient for pre- and post-PCR sample processing
  • Easy to use for pipetting steps
  • Supports 48-well and 96-well plate formats
  • Obtain uniform results with all instruments that have heated sample covers to eliminate condensation and need for mineral oil overlay
  • Proprietary MicroAmp Reaction Tubes are essential for high-performance PCR, since they ensure greatest temperature and amplification uniformity for your samples

  • Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7000 System, 7300 System, 7500 Fast Dx System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio Dx, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti™ Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA 7 Dx System, ViiA 7 Fast System, ViiA 7 System

See user's manual or package insert for limited label license and trademark information.

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 10 MicroAmp™ 96-Well Support Bases. Store at room temperature.
Packaging 10 Bases
Product Line MicroAmp
Color Black
Product Type 96-Well Plate Base
For Use With (Equipment) 2720 Thermal Cycler, 3500xL Dx Genetic Analyzer, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, AB 2720 Thermal Cycler, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, 3500xL Genetic Analyzer, QuantStudio™ Dx, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, ViiA™ 7 Fast System, 3730xl DNA Analyzer, 7500 Fast Dx System, 3130 Genetic Analyzer, 2720 Thermal Cycler, 3730 DNA Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer
No. of Wells 96
For Use With (Application) PCR
Plate Compatibility 96-Well Plates
Quantity 10 bases
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.