Learn More
AB Sciex LLC FG PROTEIN PREP KIT 100 ASSAY
Catalog No. NC1474115
Encompass
FG, KIT PROTEIN PREPARATION 100 ASSAY.Protein preparation kit containing enough reagent for the solubilization, reduction, and alkylation of 100 1-mg protein samples. The kit contains 1 vial of denaturant, 1 vial of reducing reagent, 1 vial of cysteine blocking reagent, and 6 vials of digestion buffer.
Catalog No. NC1474115 Supplier AB Sciex LLC Supplier No. 4445247
FG, KIT PROTEIN PREPARATION 100 ASSAY.Protein preparation kit containing enough reagent for the solubilization, reduction, and alkylation of 100 1-mg protein samples. The kit contains 1 vial of denaturant, 1 vial of reducing reagent, 1 vial of cysteine blocking reagent, and 6 vials of digestion buffer.