Learn More
Transforming Technologies FG6200 Series - Static Safe Cleanroom Hot Sleeves
SDP
Catalog No. 17226945
Change view
Click to view available options
Color:
Gray
Catalog No. Color
17-226-945 Gray
1 options
Catalog No. 17-226-945 Supplier Transforming Technologies Supplier No. FG6200
Add to Cart
Add to Cart
Comfortable and washable, these heat-resistant sleeves protect upper arms from elevated temperatures applications without compromising cleanliness or potential damage due to static discharge.
  • Resists Heat Up to 572°F
  • Made with Nomex™ Fabric
  • Cleanroom Compatible
  • Static Dissipative
Specifications
Size Standard
Material Aluminized Carbon, Dupont™ Kevlar™ aramid fibers
Color Gray
Style Elastic at Both Ends