Learn More
Aviva Systems Biology Corp FGA Antibody - middle region (ARP41759_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41759_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FGA. Alternate names: Fib2, MGC119422, MGC119423, MGC119425ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57986