Learn More
Aviva Systems Biology Corp FGD1 Antibody - C-terminal region (ARP31673_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31673_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FGD1. Alternate names: AAS, FGDY, MRXS16, ZFYVE3ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52834