Learn More
Aviva Systems Biology Corp FGD3 Antibody - middle region (ARP62387_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62387_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGD3. It was validated on Western Blot. . Alternate names: FLJ00004, MGC117260, ZFYVE5ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-668