Learn More
AdipoGen FGF-1 (human) (rec.) (untagged) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B01373010
Encompass_Preferred
Protein. Human FGF-1 (aa 16-155) is untagged. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. In PBS containing 0.2 mM DTT. Purity: >95% (SDS-PAGE). Fibroblast growth factors (FGFs) constitute a family of heparin-binding polypeptides involved in the regulation of biological responses such as growth, differentiation and angiogenesis. FGF-1 is a powerful mitogen of cells of mesodermal, ectodermal and endodermal origin. FGF-1 association with heparan sulfate is a prerequisite for activation of FGF receptors. FGF-1 plays a role in various stages of development and morphogenesis as well as in angiogenesis and wound healing processes. Recent data indicate a role of FGF-1 in inflammation and obesity. FGF-1 is selectively induced in fat cells by high-fat diet feeding and established the PPARgamma-FGF-1 axis as a critical pathway that regulates adipose tissue remodeling.
Catalog No. 50-205-3219