Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fibroblast growth factor 1 (FGF-1), recombinant mouse | >95.0% | 10UG
SDP

Catalog No. 5000178124
Encompass_Preferred

Recombinant mouse fibroblast growth factor 1 (FGF-1) is supplied as a lyophilized N-His-tagged protein corresponding to the mature sequence (F16-D155) and produced in E. coli for research use in cell proliferation and signaling assays.

  • Species: Mouse, mature FGF-1 sequence (F16-D155).
  • Expression system: Escherichia coli with N-terminal His tag.
  • Form: Lyophilized powder; formulated from filtered buffer with possible trehalose.
  • Purity and endotoxin: >95% by SDS-PAGE; endotoxin <1 EU/μg.
  • Molecular weight and activity: ~15.8 kDa predicted (≈17-18 kDa by SDS-PAGE); ED50 in NIH-3T3 assays ≤0.4015 ng/mL.
  • Storage and handling: store at -20°C; reconstitute at ≥100 μg/mL and aliquot for long-term storage with carrier protein.

Catalog No. 50-001-78124 Supplier Medchemexpress LLC Supplier No. HYP70028210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse fibroblast growth factor 1 (FGF-1) is supplied as a lyophilized N-His-tagged protein corresponding to the mature sequence (F16-D155) and produced in E. coli for research use in cell proliferation and signaling assays.

  • Species: Mouse, mature FGF-1 sequence (F16-D155).
  • Expression system: Escherichia coli with N-terminal His tag.
  • Form: Lyophilized powder; formulated from filtered buffer with possible trehalose.
  • Purity and endotoxin: >95% by SDS-PAGE; endotoxin <1 EU/μg.
  • Molecular weight and activity: ~15.8 kDa predicted (≈17-18 kDa by SDS-PAGE); ED50 in NIH-3T3 assays ≤0.4015 ng/mL.
  • Storage and handling: store at -20°C; reconstitute at ≥100 μg/mL and aliquot for long-term storage with carrier protein.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.