Learn More
Medchemexpress LLC FGF-1 protein, mouse (N-His) | ≥98.0% | 1MG
SDP
Supplier:  Medchemexpress LLC HYP7002821MG
Encompass_Preferred
Recombinant mouse fibroblast growth factor 1 (FGF-1) with an N-terminal His tag, expressed in E. coli and supplied as a lyophilized protein. The material is supplied with analytical characterization for purity, endotoxin, and biological activity and is intended for cell biology and biochemical research applications.
Catalog No. 50-001-77319