Learn More
Enzo Life Sciences FGF-10 (human), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2340025
Encompass_Preferred
Alternative name: Fibroblast growth factor 10, Keratinocyte growth factor 2. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium phosphate, pH 7.5. Source: Produced in E. coli. Contains 170 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4033