Learn More
Medchemexpress LLC FGF-12 Human 177 a 100ug
SDP
Catalog No. 5000180853
Encompass_Preferred
FGF-12 Protein Human (177 a a) is a single polypeptide chain with a molecular mass of 20 4 kDa FGF12 can play a role in tissues by translocating into cells through the plasma membrane
Catalog No. 50-001-80853 Supplier Medchemexpress LLC Supplier No. HYP7343100UG
FGF-12 Protein Human (177 a a) is a single polypeptide chain with a molecular mass of 20 4 kDa FGF12 can play a role in tissues by translocating into cells through the plasma membrane