Learn More
Medchemexpress LLC FGF-12 Human 181a. 100ug
SDP
Catalog No. 5000196255
Encompass_Preferred
Recombinant human FGF-12 (181 amino acids) is a His-tagged protein expressed in E. coli and supplied as a lyophilized powder for research use. It modulates neuronal voltage-gated sodium channels and is provided with analytical quality data supporting purity, molecular weight, endotoxin level, and biological activity.
  • High purity (≥95%) as determined by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, 500 mM arginine, pH 8.0 to enhance stability.
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays.
  • Demonstrated biological activity (ED50 1.702 ng/mL in NIH-3T3 assay).
  • Recommended reconstitution at ≥100 μg/mL and storage in aliquots at -20°C or -80°C.
Catalog No. 50-001-96255 Supplier Medchemexpress LLC Supplier No. HYP71959A100UG
Recombinant human FGF-12 (181 amino acids) is a His-tagged protein expressed in E. coli and supplied as a lyophilized powder for research use. It modulates neuronal voltage-gated sodium channels and is provided with analytical quality data supporting purity, molecular weight, endotoxin level, and biological activity.
  • High purity (≥95%) as determined by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, 500 mM arginine, pH 8.0 to enhance stability.
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays.
  • Demonstrated biological activity (ED50 1.702 ng/mL in NIH-3T3 assay).
  • Recommended reconstitution at ≥100 μg/mL and storage in aliquots at -20°C or -80°C.