Learn More
Medchemexpress LLC FGF-13 Human 10ug
SDP
Catalog No. 5000181543
Encompass_Preferred
FGF-13 protein is a microtubule-binding protein that directly binds and stabilizes tubulin and affects microtubule polymerization It exerts negative regulation on axonal and leading process branches which are critical for neuronal polarization and migration in the cerebral cortex and hippocampus FGF-13 Protein Human is the recombinant human-derived FGF-13 protein expressed by E coli with tag free
Catalog No. 50-001-81543 Supplier Medchemexpress LLC Supplier No. HYP7280310UG
FGF-13 protein is a microtubule-binding protein that directly binds and stabilizes tubulin and affects microtubule polymerization It exerts negative regulation on axonal and leading process branches which are critical for neuronal polarization and migration in the cerebral cortex and hippocampus FGF-13 Protein Human is the recombinant human-derived FGF-13 protein expressed by E coli with tag free