Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF-15 Mouse His-S 5ug
SDP

Supplier:  Medchemexpress LLC HYP715265UG

Encompass_Preferred

Recombinant mouse FGF-15 protein with an N-terminal His-SUMO fusion, expressed in Escherichia coli and supplied lyophilized. The preparation includes amino acids R26-K218 and is purified to >90% by reducing SDS-PAGE. Supplied in small-size formats for biochemical and cell biology research; store frozen and aliquot after reconstitution to preserve stability and activity.

  • Contains amino acids R26-K218 of mouse FGF-15.
  • N-terminal His-SUMO tag facilitates purification and improves solubility.
  • Expressed in Escherichia coli for recombinant production.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose or Tris/arginine buffers for stability.
  • Available in small sizes suitable for pilot or scale-down studies.
  • Store at -20°C; aliquot after reconstitution to minimize freeze-thaw.

Catalog No. 50-001-96754


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.