Learn More
Medchemexpress LLC HY-P7344 10ug Medchemexpress, FGF-16 (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 502083152
Encompass_Preferred
Medchemexpress, HY-P7344 10ug FGF-16 (CHO-expressed), Human CAS: Recombinant Human Fibroblast Growth Factor 16 (CHO-expressed) is a polypeptide chain containing 206 amino acids produced in CHO cells. FGF-16 is a heparin binding growth factor, a member of the FGF family. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3152 Supplier Medchemexpress LLC Supplier No. HYP734410UG
Medchemexpress, HY-P7344 10ug FGF-16 (CHO-expressed), Human CAS: Recombinant Human Fibroblast Growth Factor 16 (CHO-expressed) is a polypeptide chain containing 206 amino acids produced in CHO cells. FGF-16 is a heparin binding growth factor, a member of the FGF family. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.