Learn More
Medchemexpress LLC FGF-17 Human HEK29 10ug
SDP
Catalog No. 5000182403
Encompass_Preferred
FGF-17 Protein plays a crucial role in embryonic development serving as a signaling molecule for brain induction and patterning Its presence is vital for normal brain development emphasizing its significance in shaping embryogenesis intricacies FGF-17 interacts with FGFR3 and FGFR4 underscoring its involvement in signaling cascades that precisely orchestrate developmental events in the embryonic brain FGF-17 Protein Human (HEK293 His) is the recombinant human-derived FGF-17 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82403 Supplier Medchemexpress LLC Supplier No. HYP7265210UG
FGF-17 Protein plays a crucial role in embryonic development serving as a signaling molecule for brain induction and patterning Its presence is vital for normal brain development emphasizing its significance in shaping embryogenesis intricacies FGF-17 interacts with FGFR3 and FGFR4 underscoring its involvement in signaling cascades that precisely orchestrate developmental events in the embryonic brain FGF-17 Protein Human (HEK293 His) is the recombinant human-derived FGF-17 protein expressed by HEK293 with C-6 His labeled tag