Learn More
Medchemexpress LLC FGF-17 Mouse His 2ug
SDP
Catalog No. 5000182851
Encompass_Preferred
The FGF-17 Protein is crucial in regulating embryonic development functioning as a signaling molecule for inducing and patterning the embryonic brain It plays an essential role in normal brain development and interacts specifically with FGFR3 and FGFR4 receptors pivotal in this developmental process FGF-17 Protein Mouse (His) is the recombinant mouse-derived FGF-17 expressed by E coli with C-6 His labeled tag The total length of FGF-17 Protein Mouse (His) is 194 a a
Catalog No. 50-001-82851 Supplier Medchemexpress LLC Supplier No. HYP726532UG
The FGF-17 Protein is crucial in regulating embryonic development functioning as a signaling molecule for inducing and patterning the embryonic brain It plays an essential role in normal brain development and interacts specifically with FGFR3 and FGFR4 receptors pivotal in this developmental process FGF-17 Protein Mouse (His) is the recombinant mouse-derived FGF-17 expressed by E coli with C-6 His labeled tag The total length of FGF-17 Protein Mouse (His) is 194 a a