Learn More
Medchemexpress LLC FGF-17 Mouse His 50ug
SDP
Catalog No. 5000181410
Encompass_Preferred
The FGF-17 Protein is crucial in regulating embryonic development functioning as a signaling molecule for inducing and patterning the embryonic brain It plays an essential role in normal brain development and interacts specifically with FGFR3 and FGFR4 receptors pivotal in this developmental process FGF-17 Protein Mouse (His) is the recombinant mouse-derived FGF-17 expressed by E coli with C-6 His labeled tag The total length of FGF-17 Protein Mouse (His) is 194 a a
Catalog No. 50-001-81410 Supplier Medchemexpress LLC Supplier No. HYP7265350UG
The FGF-17 Protein is crucial in regulating embryonic development functioning as a signaling molecule for inducing and patterning the embryonic brain It plays an essential role in normal brain development and interacts specifically with FGFR3 and FGFR4 receptors pivotal in this developmental process FGF-17 Protein Mouse (His) is the recombinant mouse-derived FGF-17 expressed by E coli with C-6 His labeled tag The total length of FGF-17 Protein Mouse (His) is 194 a a