Learn More
Medchemexpress LLC HY-P7123 10ug Medchemexpress, FGF-18, Human CAS: Purity:>98%
SDP
Catalog No. 501970568
Encompass_Preferred
Medchemexpress, HY-P7123 10ug FGF-18, Human CAS: Recombinant Human Fibroblast Growth Factor 18 is a heparin-binding polypeptide growth factor, involved in cartilage growth, maturation and the development of functional cartilage and bone tissue. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0568 Supplier Medchemexpress LLC Supplier No. HYP712310UG
Medchemexpress, HY-P7123 10ug FGF-18, Human CAS: Recombinant Human Fibroblast Growth Factor 18 is a heparin-binding polypeptide growth factor, involved in cartilage growth, maturation and the development of functional cartilage and bone tissue. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.