Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z035891
Encompass
Murine FGF-18 is encoded by the FGF18 gene. By phylogenetic analysis and gene location analysis FGF-18 is divided into FGF-8 subfamily which has three members FGF-8 FGF-17 and FGF-18. Using FGF knockout mice model the numbers of this subfamily were testified that have crucial roles in embryo development. FGF-18–/– mice have decreased expression of osteogenic markers and delayed long-bone ossification. FGF-18 has been shown in vitro that this protein is able to induce neurite outgrowth in PC12 cells. In addition it also has significant roles in lung development and has an anabolic effect on cartilage formation.
Catalog No. 50-252-9599