Learn More
Medchemexpress LLC FGF-18 Rat 100ug
SDP
Catalog No. 5000196959
Encompass_Preferred
Recombinant rat FGF-18 is a heparin-binding fibroblast growth factor involved in cartilage growth, maturation, and the development and repair of bone and cartilage tissue. Supplied as a lyophilized fragment corresponding to residues E28-R199 of the rat sequence, it is intended for in vitro receptor-binding studies, cell-based activity assays, and other biochemical applications where defined FGF-18 activity is required.
  • High purity (>95%) protein suitable for biochemical and cell-based assays.
  • Expressed in Escherichia coli for consistent batch production.
  • Lyophilized formulation dialyzed against PBS or 20 mM PB, 150 mM NaCl, pH 7.4.
  • Demonstrated bioactivity with ED50 <0.5 μg/mL in 3T3 cell assays.
  • Reconstitution guidance recommends a concentration of at least 100 μg/mL to preserve activity.
  • Stable when aliquoted and stored at -20°C or -80°C for extended storage.
Catalog No. 50-001-96959 Supplier Medchemexpress LLC Supplier No. HYP7124100UG
Recombinant rat FGF-18 is a heparin-binding fibroblast growth factor involved in cartilage growth, maturation, and the development and repair of bone and cartilage tissue. Supplied as a lyophilized fragment corresponding to residues E28-R199 of the rat sequence, it is intended for in vitro receptor-binding studies, cell-based activity assays, and other biochemical applications where defined FGF-18 activity is required.
  • High purity (>95%) protein suitable for biochemical and cell-based assays.
  • Expressed in Escherichia coli for consistent batch production.
  • Lyophilized formulation dialyzed against PBS or 20 mM PB, 150 mM NaCl, pH 7.4.
  • Demonstrated bioactivity with ED50 <0.5 μg/mL in 3T3 cell assays.
  • Reconstitution guidance recommends a concentration of at least 100 μg/mL to preserve activity.
  • Stable when aliquoted and stored at -20°C or -80°C for extended storage.