Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF-19 Human 100ug
SDP

Catalog No. 5000195640
Encompass_Preferred

Recombinant human FGF-19 (mature sequence R23-K216) is a tag-free protein for in vitro research of bile acid signaling and hepatic metabolism. It engages the FGFR4/β-Klotho receptor complex to stimulate hepatic protein and glycogen synthesis via an insulin-independent endocrine pathway. Expressed in E. coli and supplied at >95% purity, it is available in research sizes including 100 μg.

  • Tag-free, full-length mature sequence for native activity.
  • Expressed in E. coli for consistent recombinant production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted molecular weight ≈21.7 kDa, compatible with SDS-PAGE analysis.
  • Available in multiple research sizes, including 100 μg, for experimental flexibility.

Catalog No. 50-001-95640 Supplier Medchemexpress LLC Supplier No. HYP7172100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human FGF-19 (mature sequence R23-K216) is a tag-free protein for in vitro research of bile acid signaling and hepatic metabolism. It engages the FGFR4/β-Klotho receptor complex to stimulate hepatic protein and glycogen synthesis via an insulin-independent endocrine pathway. Expressed in E. coli and supplied at >95% purity, it is available in research sizes including 100 μg.

  • Tag-free, full-length mature sequence for native activity.
  • Expressed in E. coli for consistent recombinant production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted molecular weight ≈21.7 kDa, compatible with SDS-PAGE analysis.
  • Available in multiple research sizes, including 100 μg, for experimental flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.