Learn More
AdipoGen FGF-2 human rec
SDP
Supplier:  AdipoGen AG40T0040C010
Encompass_Preferred
Protein. Human FGF-2 153aa. Source E. coli. Endotoxin content <0.1ng/µg protein. Lyophilized. Contains PBS. Purity >98% SDS-PAGE. FGF-2 plays an important role in the regulation of angiogenesis, cell survival, cell division, wound healing and tissue repair, embryonic development, cell differentiation and cell migration, neuronal function and neural degeneration. It functions as potent mitogen in vitro.
Catalog No. 50-256-2791