Learn More
AdipoGen FGF-2 human rec
SDP
Catalog No. 502562790
Encompass_Preferred
Protein. Human FGF-2 153aa. Source E. coli. Endotoxin content <0.1ng/µg protein. Lyophilized. Contains PBS. Purity >98% SDS-PAGE. FGF-2 plays an important role in the regulation of angiogenesis, cell survival, cell division, wound healing and tissue repair, embryonic development, cell differentiation and cell migration, neuronal function and neural degeneration. It functions as potent mitogen in vitro.
Catalog No. 50-256-2790 Supplier AdipoGen Supplier No. AG40T0040C050
Protein. Human FGF-2 153aa. Source E. coli. Endotoxin content <0.1ng/µg protein. Lyophilized. Contains PBS. Purity >98% SDS-PAGE. FGF-2 plays an important role in the regulation of angiogenesis, cell survival, cell division, wound healing and tissue repair, embryonic development, cell differentiation and cell migration, neuronal function and neural degeneration. It functions as potent mitogen in vitro.