Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF-2 Mouse 145a.a 50ug
SDP

Catalog No. 5000185013
Encompass_Preferred

Recombinant mouse fibroblast growth factor 2 (FGF-2) comprises the core 145-amino-acid sequence, supplied lyophilized and tag free. Produced in E. coli and characterized by high purity, this protein is intended for cell signaling, proliferation, and tissue repair research.

  • Consists of 145 amino acids with a predicted mass of 16.3 kDa.
  • Expressed in Escherichia coli and provided tag free.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 for stability.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or frozen for long-term storage.
  • Suitable for cell signaling, proliferation, and tissue repair assays.

Catalog No. 50-001-85013 Supplier Medchemexpress LLC Supplier No. HYP706650UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse fibroblast growth factor 2 (FGF-2) comprises the core 145-amino-acid sequence, supplied lyophilized and tag free. Produced in E. coli and characterized by high purity, this protein is intended for cell signaling, proliferation, and tissue repair research.

  • Consists of 145 amino acids with a predicted mass of 16.3 kDa.
  • Expressed in Escherichia coli and provided tag free.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 for stability.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or frozen for long-term storage.
  • Suitable for cell signaling, proliferation, and tissue repair assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.