Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGF-2 Mouse 145a.a 500ug
SDP

Catalog No. 5000185308
Encompass_Preferred

Recombinant mouse FGF-2 (145 amino acids) is a tag-free, lyophilized growth factor produced in E. coli and supplied as a 500 μg vial. It activates FGFR-mediated signaling to promote cell proliferation, angiogenesis, and cartilage and bone repair, and is provided at >95% purity for research applications.

  • Contains 145 amino acids, predicted mass 16.3 kDa.
  • Produced in E. coli and presented tag free.
  • Lyophilized from PBS (pH 7.4) for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Supplied as a 500 μg amount for experimental use.
  • Store at -20°C; reconstituted material stable at 4°C for 1 week.

Catalog No. 50-001-85308 Supplier Medchemexpress LLC Supplier No. HYP7066500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse FGF-2 (145 amino acids) is a tag-free, lyophilized growth factor produced in E. coli and supplied as a 500 μg vial. It activates FGFR-mediated signaling to promote cell proliferation, angiogenesis, and cartilage and bone repair, and is provided at >95% purity for research applications.

  • Contains 145 amino acids, predicted mass 16.3 kDa.
  • Produced in E. coli and presented tag free.
  • Lyophilized from PBS (pH 7.4) for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Supplied as a 500 μg amount for experimental use.
  • Store at -20°C; reconstituted material stable at 4°C for 1 week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.