Learn More
Medchemexpress LLC FGF-2 Mouse His 100ug
SDP
Catalog No. 5000180936
Encompass_Preferred
FGF-2 is a member of the fibroblast family involved in bone healing cartilage repair bone repair and nerve regeneration FGF-2 is also a mitotic promoter that accelerates cell proliferation FGF-2 regulates immune processes by specifically targeting tyrosine kinase receptors and activating the FGF/FGFR signaling pathway For example FGF-2 is involved in the JAK-STAT signaling pathway to regulate cartilage metabolism and also activates ERK signaling to promote cartilage regeneration FGF-2 combined with FGFR1/3 promoted degeneration and repair of articular cartilage respectively[1] FGF-2 is also a known carcinogen in GBM which contributes to glioma growth and vascularization[2] FGF-2 Protein Mouse (His) consists of 144 amino acids produced by E coli with N-His
Catalog No. 50-001-80936 Supplier Medchemexpress LLC Supplier No. HYP73052100UG
FGF-2 is a member of the fibroblast family involved in bone healing cartilage repair bone repair and nerve regeneration FGF-2 is also a mitotic promoter that accelerates cell proliferation FGF-2 regulates immune processes by specifically targeting tyrosine kinase receptors and activating the FGF/FGFR signaling pathway For example FGF-2 is involved in the JAK-STAT signaling pathway to regulate cartilage metabolism and also activates ERK signaling to promote cartilage regeneration FGF-2 combined with FGFR1/3 promoted degeneration and repair of articular cartilage respectively[1] FGF-2 is also a known carcinogen in GBM which contributes to glioma growth and vascularization[2] FGF-2 Protein Mouse (His) consists of 144 amino acids produced by E coli with N-His