Learn More
Medchemexpress LLC FGF-21 Cynomolgus 10ug
SDP
Supplier:  Medchemexpress LLC HYP7265010UG
Encompass_Preferred
FGF-21 is a metabolic regulator and a potential anti-diabetic agent FGF-21 regulates energy balance and glucose and lipid homeostasis through a heterodimeric receptor complex comprising FGFR1 and -klotho FGF-21 can also signal through FGFR2 and FGFR3 FGF-21 can be used for research of obesity NASH NAFLD and type 2 diabetes mellitus[1][2] FGF-21 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived FGF-21 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-81408