Learn More
Medchemexpress LLC FGF-21 Cynomolgus 50ug
SDP
Catalog No. 5000181409
Encompass_Preferred
FGF-21 is a metabolic regulator and a potential anti-diabetic agent FGF-21 regulates energy balance and glucose and lipid homeostasis through a heterodimeric receptor complex comprising FGFR1 and -klotho FGF-21 can also signal through FGFR2 and FGFR3 FGF-21 can be used for research of obesity NASH NAFLD and type 2 diabetes mellitus[1][2] FGF-21 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived FGF-21 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-81409 Supplier Medchemexpress LLC Supplier No. HYP7265050UG
FGF-21 is a metabolic regulator and a potential anti-diabetic agent FGF-21 regulates energy balance and glucose and lipid homeostasis through a heterodimeric receptor complex comprising FGFR1 and -klotho FGF-21 can also signal through FGFR2 and FGFR3 FGF-21 can be used for research of obesity NASH NAFLD and type 2 diabetes mellitus[1][2] FGF-21 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived FGF-21 protein expressed by HEK293 with C-6 His labeled tag