Learn More
AdipoGen FGF-21 hFc h rec
SDP
Catalog No. 502563002
Encompass_Preferred
Protein. Human FGF-21 aa 1-209 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >95% SDS-PAGE. FGF-21 Fibroblast growth factor 21 stimulates glucose uptake in differentiated adipocytes via the induction of glucose transporter SLC2A1/GLUT1 expression but not SLC2A4/GLUT4 expression. Activity requires the presence of betaKlotho.
Catalog No. 50-256-3002 Supplier AdipoGen Supplier No. AG40A0095C010
Protein. Human FGF-21 aa 1-209 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >95% SDS-PAGE. FGF-21 Fibroblast growth factor 21 stimulates glucose uptake in differentiated adipocytes via the induction of glucose transporter SLC2A1/GLUT1 expression but not SLC2A4/GLUT4 expression. Activity requires the presence of betaKlotho.