Learn More
AdipoGen FGF-21 (human) (rec.) (10 µg)
SDP
Supplier:  AdipoGen AG40A0091YC010
Encompass_Preferred
Protein. Human FGF-21 (aa 29-209) is fused at the C-terminus to a FLAG-tag. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Liquid. Contains PBS. Purity: >90% (SDS-PAGE). FGF-21 (Fibroblast growth factor 21) stimulates glucose uptake in differentiated adipocytes via the induction of glucose transporter SLC2A1/GLUT1 expression (but not SLC2A4/GLUT4 expression). Activity requires the presence of betaKlotho.
Catalog No. 50-205-3060